Learn More
Honeywell™ GRIP Latex Gloves
Catalog Number p-7159518
Click to view available options
Size:
9
10
Honeywell™ GRIP Latex Gloves for handling of heavy and slightly sharp parts in a damp or non greasy environment.
  • Natural latex coated glove on cotton jersey base
  • Crinkled finishing
  • Knitted wrist
  • The cotton absorbs perspiration thus provides greater comfort
  • The ventilated back improves the glove's breathability
  • The glove's crinkled latex coating provides resistance to abrasion and perforation as well as an excellent grip
  • Natural latex provides excellent flexibility, synonymous with dexterity and comfort
  • It also offers good abrasion and tear resistance
  • Its crepe finish helps the user to grasp slippery parts
TRUSTED_SUSTAINABILITY
Certifications/Compliance EN 420-2003 Basic standard, EN 388-2016 Mechanical risks
Coating Material Latex
Exterior Finish Crinkle
Material Cotton
Size 9
Length (Metric) 26 cm