missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Grey Natural Rubber Filter Ring with Flange

Brand:  Deutsch Neumann™ 2350011

Code : A0

Additional Details : Weight : 0.08300kg

 View more versions of this product

Product Code. 10241941

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Gray
1Each
41 mm
Natural Rubber
1 Each
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers