missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Grey Natural Rubber Filter Ring with Flange
Brand: Deutsch Neumann™ 2350021
Code : A0
Additional Details : Weight : 0.80000kg
Specifications
Gray | |
1Each | |
48 mm |
Natural Rubber | |
1 Each | |
20 mm |