Learn More
Kimberly-Clark™ Professional WYPALL™ Cleaning Wiper Refill, Green
Cleans and protects with a rough side to scrub off dirt and a smooth side to clean and finish. Kimberly-Clark™ WYPALL™ Cleaning Wipers Refill is supplied pre-soaked.
Brand: Kimberly-Clark™ Professional 7776
Code : 02
Additional Details : Weight : 0.01000kg
Description
- Green, pre-soaked wipers
- Rough side scrubs while smooth side polishes
- Ideal for cleaning hands, tools, machinery without soap or water
- Protects hands and reduces skin dryness
- Refills for use with WYPALL™ Cleaning Wiper Dispenser
Specifications
270mm | |
Green | |
For cleaning hands, tools, machinery without soap or water, protecting hands, reducing skin dryness, improving hygiene | |
270mm | |
6 bags x 75 sheets |
4.672kg (10.29 lbs._ | |
270 x 270mm | |
10.63 in. | |
Wypall Cleaning Wipe Refill |