missing translation for 'onlineSavingsMsg'
Learn More

Key Industrial Equipment™ Gravity roller bearing conveyor, 70kg steel roller bearing capacity, width 750mm, pitch 150mm

Brand:  Key Industrial Equipment™ A341746

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12831489

  • £168.84 / Each

Estimated Shipment date
from Supplier 24-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Conveyor tracks manufactured from high quality 2mm thick steel and supplied complete with zinc plated steel or high impact PVC roller bearings. Height adjustable supports allow track to decline for smooth movement. Low maintenance for trouble free service. Track sides are pre-punched every 150mm to accept support stands and guard rails.
  • Roller bearing pitch: 150mm
  • Track width: 750mm
Videos
Documents