missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Gravity roller bearing conveyor, 70kg steel roller bearing capacity, width 450mm, pitch 150mm
Brand: Manutan A341748
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Conveyor tracks manufactured from high quality 2mm thick steel and supplied complete with zinc plated steel or high impact PVC roller bearings. Height adjustable supports allow track to decline for smooth movement. Low maintenance for trouble free service. Track sides are pre-punched every 150mm to accept support stands and guard rails.- Roller bearing pitch: 150mm
- Track width: 450mm