Learn More
Fisherbrand™ Gravity Convection Microbiological Incubator
Ideal incubator for basic incubation applications and heated storage
£1060.00 - £2075.00
Specifications
Convection Type Gravity
Display Digital
Material Stainless steel
No. of Shelves Two
Plug Type UK and EU
View More Specs
Products 3
Product Code Brand Capacity (Metric) Dimensions (L x W x H) Exterior Price Quantity & Availability  
Product Code Brand Capacity (Metric) Dimensions (L x W x H) Exterior Price Quantity & Availability  
15835911 Fisherbrand™
194L 738 x 640 x 920mm £2075.00
Each View alternative products
Save Up to 
»
In Stock 1
Add to basket
15815911 Fisherbrand™
117L 565 x 640 x 820mm £1490.00
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
15895901 Fisherbrand™
75L 565 x 530 x 720mm £1060.00
Each View alternative products
Save Up to 
»
In Stock 1
Add to basket
Description
Description
  • Ideal for microbiological incubation and heated storage
  • Maximum temperature 75°C
  • Single set point digital control
  • Inner glass door for minimal disturbance while viewing samples
  • Stackable, no tools or kit required
  • Access port standard
  • Stainless steel interior
  • Two year parts and labor warranty
  • CE marked
  • Two perforated stainless steel shelves included as standard
  • Extra shelves also available
Specifications
Specifications
Gravity
Stainless steel
UK and EU
Ambient +7 to 75°C
Digital
Two
230V, 50/60Hz
Two year parts and labor warranty