missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Graphite tape, square weave, 0.56mm (0.022 in.) thick x 25mm (0.98in) wide, 99.9%

Brand:  Alfa Aesar™ 010138.BW

Code : NEW

Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg

Quantity 50cm

 View more versions of this product

Product Code. 11391188

  • £54.30 / 50cm


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Graphite tape
Square weave, 0.56mm (0.022 in.) thick x 25mm (0.98 in.) wide
MFCD00144065

Specifications

Graphite tape
Square weave, 0.56mm (0.022 in.) thick x 25mm (0.98 in.) wide
MFCD00144065
50cm
99.9%
Videos
SDS