missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Graphite tape, square weave, 0.56mm (0.022 in.) thick x 25mm (0.98in) wide, 99.9%

Brand:  Thermo Scientific Alfa Aesar 010138.CB

Graphite tape, square weave, 0.56mm (0.022 in.) thick x 25mm (0.98in) wide, 99.9%

Code : NEW

Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg

Product Code. 11301198

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.