Learn More
Graphite rod, pyrolytic coated, 2mm (0.08in) dia x 152mm (6in) long, Thermo Scientific Chemicals
£90.50 - £364.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436545
|
Thermo Scientific Alfa Aesar
043670.KG |
1 Pc. |
£90.50
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15446545
|
Thermo Scientific Alfa Aesar
043670.KJ |
5 Pc. |
£364.00
Pack of 5 |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Graphite rod, is suitable for making: Anodes, Electrodes, Heating elements, Crucibles, Molds & Ingots, Hot press dies, Gas nozzles, Lubrication sticks, Bushings & bearings.