missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Graphite rod, 6.3mm (0.25 in.) dia.. x 61cm (24 in.) long, 99% (metals basis)

Brand:  Alfa Aesar™ 010135.KI

Code : NEW

Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg

 View more versions of this product

Product Code. 11315749

  • £39.40 / Pack of 4


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Specifications

Graphite rod
4 pieces
Pieces
(metals basis)
≈40g/pc
6.3mm (0.25 in.) dia. x 61cm (24 in.) long
99%
MFCD00144065
Videos
SDS