missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Graphite rod, 6.15mm (0.242 in.) dia. x 102mm (4 in.) long, 99.9995% (metals basis)

Brand:  Alfa Aesar™ 040766.KD

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15493725

  • £146.90 / Pack of 50


EU Stock 3
Add to basket

Specifications

Specifications

Graphite rod
6.15mm (0.242 in.) dia. x 102mm (4 in.) long
Rod
(metals basis)
50 pieces
AGKSP grade, ultra 'F' purity
99.9995%
MFCD00144065
Videos
SDS