missing translation for 'onlineSavingsMsg'
Learn More

Graphite powder, natural, high purity, -200 mesh, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15416055
Click to view available options
Quantity:
28 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

Graphite powder is used to make crucibles, lead pencils, electrodes, lubricants, stove polish, brake linings, batteries, glass manufacture, conductive coatings, and for foundry facings. It is used in the synthesis of single sheet graphene nanoflakes and graphite based magnetorheological elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7782-42-5
Mesh Size −200 Mesh
Odor Odorless
Molecular Formula C
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
Quantity 28 g
UN Number UN1325
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
Solubility Information Insoluble in water.
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.01
PubChem CID 5462310
ChEBI CHEBI:27594
Percent Purity 99.9999%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Graphite powder, Natural
Grade Ultra superior purity
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'