Learn More
Granzyme B Monoclonal Antibody (NGZB), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 25-8898-82
175.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: This NGZB monoclonal antibody reacts with mouse Granzyme B, which is a member of the granzyme serine protease family. Granzyme B is found in the granules of cytotoxic T cells and NK cells. Granzyme B has also been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved has been identified as mannose 6-phosphate receptor. This receptor functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. This NGZB monoclonal antibody does not crossreact to human Granzyme B nor is staining blocked with GB11, suggesting it recognizes a different epitope. Applications Reported: This NGZB antibody has been reported for use in intracellular staining and flow cytometric analysis. Applications Tested: This NGZB antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins. This can be used at less than or equal to 0.
Granzyme B Monoclonal antibody specifically detects Granzyme B in Mouse samples. It is validated for Flow Cytometry.Specifications
Granzyme B | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P04187 | |
GZMB | |
Affinity chromatography | |
RUO | |
14939 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
NGZB | |
PE-Cyanine7 | |
GZMB | |
AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.