missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Monoclonal Antibody (GB11), PE-Cyanine5.5, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen GRB18
302.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Flow Cytometry.Specifications
Granzyme B | |
Monoclonal | |
PE-Cyanine5.5 | |
GZMB | |
AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E | |
Mouse | |
purified | |
RUO | |
3002 | |
4° C | |
Liquid |
Flow Cytometry | |
GB11 | |
PBS with 0.09% sodium azide; pH 7.2 | |
P10144 | |
GZMB | |
Human Granzyme B. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |