missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Mouse anti-Human, eFluor™ 450, Clone: N4TL33, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 48-8896-42
Additional Details : Weight : 0.01000kg
Description
Mouse Monoclonal Antibody
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Flow CytometrySpecifications
Granzyme B | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P10144 | |
AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; Cytotoxic cell protease 1; Cytotoxic T-lymphocyte proteinase 2; Fragmentin-2; Granzyme B; Granzyme B(G,H); Granzyme-2; GranzymeB; GRB; GZB; Gzmb; HLP; Human lymphocyte protein; Lymphocyte protease; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E | |
Mouse | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
N4TL33 | |
eFluor 450 | |
GZMB | |
Liquid | |
GZMB | |
IgG2a, kappa | |
100 Tests | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
3002 |