Learn More
Granzyme A Monoclonal Antibody (GzA-3G8.5), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-5831-82
180.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: This Gza-3G8.5 monoclonal antibody reacts with mouse Granzyme A. Granzyme A is the most abundantly expressed of the ten granzyme serine proteases that have been identified in mice. Granzymes are proteins released from the granules of NK cells and cytotoxic T lymphocytes that induce death in target cells by cleavage of intracellular substrates and play a critical role in immune defense against viruses, tumors, and intracellular bacteria. Granzyme A activates caspase-independent cell death pathways morphologically similar to apoptosis and characterized by mitochondrial and DNA damage. It may also play a role in inflammation, as the precursor form of IL-1 beta (pro-IL-1 beta) is among its target substrates. Granzyme A shares overlapping substrate specificity with the closely-related Granzyme K, which is believed to account for the minimal decrease in cytotoxicity of Granzyme A-deficient CTLs. Applications Reported: This GzA-3G8.5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This GzA-3G8.5 antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Granzyme A Monoclonal antibody specifically detects Granzyme A in Mouse samples. It is validated for Flow Cytometry.Specifications
Granzyme A | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P11032 | |
GZMA | |
Affinity chromatography | |
RUO | |
14938 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
GzA-3G8.5 | |
PerCP-eFluor 710 | |
GZMA | |
Autocrine thymic lymphoma granzyme-like serine protease; AW494114; BLT esterase; CTL tryptase; CTLA3; CTLA-3; Cytotoxic T-lymphocyte proteinase 1; Cytotoxic T-lymphocyte-associated serine esterase-3; fragmentin-1; granzyme 1; granzyme A; Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factor serine protease); granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3); Granzyme-1; GZMA; H factor; Hanukah factor; Hanukah factor serine protease); Hanukkah factor; Hf; HFSP; Mtsp-1; SE1; serine esterase 1; t cell-specific serine protease 1; TSP1; TSP-1 | |
Mouse | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |
For Research Use Only.