missing translation for 'onlineSavingsMsg'
Learn More

Granzyme A Mouse anti-Mouse, PE, Clone: GzA-3G8.5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5831-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15597026

  • £79.25 / 25µg

Estimated Shipment
27-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface ′nonself′ antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein described here is a T cell- and natural killer cell-specific serine protease that may function as a common component necessary for lysis of target cells by cytotoxic T lymphocytes and natural killer cells.
Specifications

Specifications

Granzyme A
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P11032
CTLA3, HFSP
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
GzA-3G8.5
PE
Gzma
Liquid
Gzma
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14938
Videos
Documents

For Research Use Only.