Learn More
Kartell™ Graduated Conical Centrifuge Tubes
Manufactured from highly translucent polypropylene for  enhanced visibility and good chemical resistance. Autoclavable, rimmed Kartell™ Cylindrical Centrifuge Tubes deliver stabilitiy in a properly balanced rotor and are suitable for use as a conventional test tube.
Brand:  Kartell™ 0230200
Code : A0
Additional Details : Weight : 0.47700kg
Product Code. 10244801
  • £64.55 / Pack of 100
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
  • Solid polypropylene construction translates to safety and economy with reduced breakage in the lab
  • Withstands temperatures up to 120°C
  • Rimmed
  • Molded graduations for reproducibility 
  • Autoclavable
  • Doubles as a conventional laboratory test tube*
*Cannot be used with naked flame