missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Grade C 160 Smooth Qualitative Filter Papers
Filters with increased wet strength for the filtration and absorption of liquids.
Brand: Sartorius™ FT-3-343-047
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Filters with increased wet strength for the filtration and absorption of liquids.Specifications
25mbar | |
Refined Pulp | |
C 160 | |
0.3 mm | |
47 mm |
Disc | |
Used for the filtration of cooking and transformer oils, galvanic baths and as base paper for further impregnation with certain reagents | |
50/Pk. | |
Qualitative Filter Boards |