Sartorius™ Grade C 160 Smooth Qualitative Filter Papers


Overview
Brand: Sartorius™

Manufacturer Part Number: FT-3-343-185

UNSPSC: 41120000

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11710486

Quantity Price
1 £ 11.0 / Pack of 50
Estimated Shipment
08-05-2017
Add to basket


Description and Specification

Specification

Thickness (Metric) 0.3mm
Diameter (Metric) 185mm
Format Filter discs
Size (Metric) 185mm
Grade C 160
Quantity 50/Pk.
Type Qualitative Filter Boards
Material Refined pulp
For Use With (Application) Used for the filtration of cooking and transformer oils, galvanic baths and as base paper for further impregnation with certain reagents
Airflow Rate 25mbar