missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 601 General Purpose Filter Papers

A filter paper used in analytical works in laboratories with medium filtration velocity.

Brand:  Fisherbrand™

Code : 47

Additional Details : Weight : 0.05850kg

 View more versions of this product

Product Code. 11582003

  • £15.60 / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

Applications:
  • Used in hospitals, research laboratories, schools, chemical industries, pharmaceutical, cosmetic, food industries
Specifications

Specifications

Disc
5 to 13 μm
General works, hospitals, research laboratories, schools, chemical industries, cosmetics, food and beverage industries
100/Pk.
85 mm
Cellulose
5 to 13 μm
601
0.16 mm
85 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers