missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Grade 541 Cotton Linter Filter Paper

Brand:  Whatman™ 1541-917

Code : 50

Additional Details : Weight : 1.94000kg

Product Code. 11310524

  • £433.60 / Pack of 100


Estimated Shipment
06-06-2018
Add to basket

Description

Description

These are the supreme quantitative filter papers featuring high wet strength and chemical resistance. These papers are acid-hardened, which reduces ash to an extremely low level. The tough surfaces of these fillters make them suitable for a wide range of critical analytical filtration operations. Each grade offers a convenient combination of filtration speed and particle retention. Grade 541: 22μm. Fast filtration of coarse particles and gelatinous precipitates in acid/alkali solutions during gravimetric analysis. Typical applications include determining fibre in animal foodstuffs, gelatin in milk and cream, chloride in cement, and chloride and phosphorous in coal and coke. Hardened ashless: 0.006% ash maximum. These filters are acid-hardened to give high wet strength and chemical resistance with extremely low ash content. The tough surface makes these filters suitable for a wide range of critical filtration procedures.
Specifications

Specifications

Filter Paper
Grade 541
Sheet
For wide range of critical analytical filtration operations
Fast
3.8 sec./100mL/sq in.
155μm
100Pack
Cotton linter
155μm
22μm
460 × 570mm
----
Videos