missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Grade 4b, Smooth Paper for Qualitative and Technical Analyses, Sheets

Sartorius Stedim Biotech™ Grade 4b, Smooth Paper for Qualitative and Technical Analyses, Sheets are suitable for analytical and technical purposes and feature medium fast filtering.

Brand:  Sartorius Stedim Biotech™ FT-2-309-193290Q

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15962750

  • £112.00 / Pack of 500

Estimated Shipment: 01-02-2022
Add to basket

Description

Description

  • Made of cotton linters and refined pulp with > 95% alpha-cellulose
  • Medium fast filtering
  • Non-sterile and bulk packed
Specifications

Specifications

Sheet
Cotton Linters and Refined Pulp (>95% alpha-cellulose)
163 x 290 mm
75 g/m2
8 to 12μm
Qualitative Filter Paper
4 b
500/Pk.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers