missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 470 Folded Filter Cellulose and Diatomaceous Earth Filter Papers

Offers better separating capability and quickly retains the finest particles at high flow rates.

Brand:  Sartorius™ FT-4-606-185

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10286131

  • £131.00 / Pack of 100
Estimated Shipment: 19-04-2024
to see stock.
Add to basket

Description

Description

  • Used for the filtration of the finest, semi-colloidal precipitates, e.g. those consisting of proteins, clay or cold-precipitated barium
  • For clarification of beer, wine, urine during spectophotometric or refractometric test
Specifications

Specifications

Folded
80sec.
470
0.32 mm
185 mm
Cellulose, Diatomaceous Earth
Offer a much better separating capability than pure cellulose papers at the same rate of filtration
100/Pk.
Diatomaceous Earth Filter Paper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers