missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Grade 392 Quantitative Papers, Folded

Sartorius Stedim Biotech™ Grade 392 Quantitative Papers, Folded are suitable for quantitative and gravimetric analyses as well as for pressure or vacuum filtration.

Brand:  Sartorius Stedim Biotech™ FT-4-105-110

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15952980

  • £41.00 / Pack of 100

Estimated Shipment: 27-01-2022
Add to basket

Description

Description

  • Made of 100% cotton linters with an alpha-cellulose content of < 98%
  • Acid-washed to make the papers ashless and achieve high purity (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
  • Medium dense, medium fast filtering
Specifications

Specifications

Folded
100% Cotton Linters (>98% alpha-cellulose; ash-free)
0.17 mm
84 g/m2
5 to 8μm
110 mm
392
100/Pk.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers