Learn More
Sartorius Stedim Biotech™ Grade 292 Qualitative Papers, Discs
Sartorius Stedim Biotech™ Grade 292 Qualitative Papers, Discs are especially suitable for qualitative analyses that require a low ash content.
Brand: Sartorius Stedim Biotech™ FT-3-205-085
Additional Details : Weight : 0.04900kg
Description
- Pure cotton linters or cotton linters with refined pulp
- No additives, such as wet-strengthening agents
Specifications
87 g/m2 | |
Disc | |
5 to 8μm | |
0.18 m |
85 mm | |
292 | |
100/Pk. | |
Cotton Linters (Low-nitrogen and Nitrates; < 0.06% ash) |