missing translation for 'onlineSavingsMsg'
Learn More

Grace™ VisionHT™ Solvent Miser Silica HPLC Columns, 3μm Particle Size

Brand:  Grace™ 5152090

Code : 16

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 12121390

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Solvent Miser
2.1mm
3μm
150mm
3μm
120Å
HPLC Column
HPLC
VisionHT™ HILIC
No
Very polar compounds with high organic mobile phases for improved sensitivity by MS
VisionHT™ HILIC
2 to 8
220m2/g
L3
VisionHT™ HILIC
Videos