missing translation for 'onlineSavingsMsg'
Learn More

Grace™ VisionHT™ Solvent Miser C18 High-Load HPLC Columns, 3μm Particle Size

Brand:  Grace™ 5151979

Code : 16

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 12191300

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

11%
VisionHT™ C18 HighLoad
Yes
General purpose for broad range of compounds. Classic selectivity, high capacity for hydrophobic compounds
3μm
120Å
HPLC Column
HPLC
Solvent Miser
2.1mm
3μm
150mm
1 to 10
220m2/g
L1
VisionHT™ C18 HighLoad
Videos