missing translation for 'onlineSavingsMsg'
Learn More

Grace™ VisionHT™ Silica HPLC Columns, 3μm Particle Size

Brand:  Grace™ 5152112

Code : 16

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 12181390

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Traditional
4.6mm
3μm
150mm
3μm
120Å
HPLC Column
HPLC
VisionHT™ Silica
No
Isomeric separation of non-aqueous compatible compounds by absorption chromatography
VisionHT™ Silica
2 to 8
220m2/g
L3
VisionHT™ Silica
Videos