Learn More
Grace™ VisionHT™ Rocket Silica HPLC Columns
High-speed, high-resolution columns for high-throughput analysis
Brand:  Grace™ 5152093
Code : 16
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 12151390
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Rocket™
7mm
3μm
53mm
3μm
120Å
HPLC Column
HPLC
VisionHT™ HILIC
No
Very polar compounds with high organic mobile phases for improved sensitivity by MS
VisionHT™ HILIC
2 to 8
220m2/g
L3
VisionHT™ HILIC
Videos