missing translation for 'onlineSavingsMsg'
Learn More

Grace™ VisionHT™ Rocket C18 Basic HPLC Columns

High-speed, high-resolution columns for high-throughput analysis

Brand:  Grace™ 5152005

Code : 16

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 12141330

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

5%
VisionHT™ C18 Basic
1.5μm
1.5μm
120Å
HPLC Column
HPLC
Rocket™
7mm
53mm
1 to 10
220m2/g
L1
VisionHT™ C18 Basic