missing translation for 'onlineSavingsMsg'
Learn More

Grace™ Jones Genesis™ C18 Reversed Phase HPLC Columns

A unique proprietary end-capping reagent provides freedom from residual silanols and enhanced stability under low-pH operating conditions

Brand:  Grace™ 5109783

Code : Z1

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10439824

  • £1004.00 / Each


Estimated Shipment
13-06-2018
Add to basket

Description

Description

  • Excellent peak symmetry
  • Exceptional stability from 1 to 10pH
  • Reduced need for mobile-phase modifiers
  • Long column life
  • C18 bonding is monomeric

Specifications

Specifications

0.8mL/min.
Silica
Spherical
HPLC Column
L1
18%
120Å
4μm
Analysis of a wide range of analytes and impressive peak symmetry for both Acidic and basic compounds
Isocratic, 50% ACN in Water
1 to 10
300m2/g
C18
250mm
Yes
4.6mm
Videos