Learn More
Grace™ Jones Genesis™ C18 Reversed Phase HPLC Columns
A unique proprietary end-capping reagent provides freedom from residual silanols and enhanced stability under low-pH operating conditions
Brand: Grace™ 5140908
Code : Z1
Additional Details : Weight : 1.00000kg
Description
- Excellent peak symmetry
- Exceptional stability from 1 to 10pH
- Reduced need for mobile-phase modifiers
- Long column life
- C18 bonding is monomeric
Specifications
4.6 mm | |
150 mm | |
C18 |
Analysis of a Wide Range of Analytes and Impressive Peak Symmetry for Both Acidic and Basic Compounds | |
3 μm | |
Column |