Grace™ GraceResolv™ Silica Flash Cartridges
High resolution and outstanding loading capacity
Overview
Brand: Grace™
Manufacturer Part Number: 5155971
UNSPSC: 41115700
Code: 50
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 12101420
Quantity Price
1 £ 943.04 / Pack of 4
Estimated Shipment
29-03-2017
Add to basket
Description and Specification
Specification
Material Silica
Bed Weight (Metric) 750g
For Use With (Equipment) For flash instruments, Isco™ and Biotage™
Resolution High
Product Type Flash cartridge
Documentation