missing translation for 'onlineSavingsMsg'
Learn More

Grace™ Alltech™ GracePure™ SPE Columns

Clean up and concentrate problematic samples

Brand:  Grace™ 5138777

Code : 16

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12121470

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Silica
1mL
SPE Column
60Å
Silica
50μm
100mg
Normal Phase
For the cleanup, concentration and recovery of chromatography samples
Silica
Silica
Videos
Documentation