missing translation for 'onlineSavingsMsg'
Learn More

GR 89696 fumarate, Tocris Bioscience™

Subtype-selective κ agonist

Brand:  Tocris (Bio-Techne) 148350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13469343

  • £454.23 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly potent and selective κ-opioid agonist (IC50 = 0.04 nM) that may be selective for the putative κ2 receptor. Anti-nociceptive and neuroprotective in vivo.
Specifications

Chemical Identifiers

126766-32-3
530.399
gr 89696 fumarate, gr-89696 fumarate, gr 89696 fumarate salt, 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylic acid methyl ester fumarate, 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylic acid methyl ester fumarate salt, 1-3,4-dichlorophenyl acetyl-4-acetyl-2-1-pyrrolidinylmethyl piperazine, methyl 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylate fumarate, gr 89696 fumarate salt, solid, 4-3,4-dichlorophenyl acetyl-3
(E)-but-2-enedioic acid;methyl 4-[2-(3,4-dichlorophenyl)acetyl]-3-(pyrrolidin-1-ylmethyl)piperazine-1-carboxylate
C23H29Cl2N3O7
ABTNETSDXZBJTE-WLHGVMLRSA-N
6442840
COC(=O)N1CCN(C(C1)CN2CCCC2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.C(=CC(=O)O)C(=O)O

Specifications

50mg
126766-32-3
gr 89696 fumarate, gr-89696 fumarate, gr 89696 fumarate salt, 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylic acid methyl ester fumarate, 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylic acid methyl ester fumarate salt, 1-3,4-dichlorophenyl acetyl-4-acetyl-2-1-pyrrolidinylmethyl piperazine, methyl 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylate fumarate, gr 89696 fumarate salt, solid, 4-3,4-dichlorophenyl acetyl-3
COC(=O)N1CCN(C(C1)CN2CCCC2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.C(=CC(=O)O)C(=O)O
530.399
530.4
GR 89696 fumarate
C23H29Cl2N3O7
ABTNETSDXZBJTE-WLHGVMLRSA-N
(E)-but-2-enedioic acid;methyl 4-[2-(3,4-dichlorophenyl)acetyl]-3-(pyrrolidin-1-ylmethyl)piperazine-1-carboxylate
6442840
>99%
Videos
Documents