missing translation for 'onlineSavingsMsg'
Learn More

GR 55562 dihydrochloride, Tocris Bioscience™

5-HT1B antagonist

Brand:  Tocris (Bio-Techne) 105450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488393

  • £582.37 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

A selective competitive 5-HT1B (5-HT1Dβ) silent antagonist with pKB values of 7.3 and 6.3 for human cloned 5-HT1B and 5-HT1D receptors respectively and only weak binding at a number of other 5-HT subtypes.
Specifications

Chemical Identifiers

159533-25-2
448.388
3-[3-(Dimethylamino)propyl]-4-hydroxy-N-[4-(4-pyridinyl)phenyl]benzamide dihydrochloride
C23H27Cl2N3O2
KBKWJHYQFQONBJ-UHFFFAOYSA-N
56972159

Specifications

GR 55562 dihydrochloride
159533-25-2
C23H27Cl2N3O2
KBKWJHYQFQONBJ-UHFFFAOYSA-N
3-[3-(dimethylamino)propyl]-4-hydroxy-N-(4-pyridin-4-ylphenyl)benzamide;dihydrochloride
56972159
50mg
>98%
3-[3-(Dimethylamino)propyl]-4-hydroxy-N-[4-(4-pyridinyl)phenyl]benzamide dihydrochloride
CN(C)CCCC1=C(C=CC(=C1)C(=O)NC2=CC=C(C=C2)C3=CC=NC=C3)O.Cl.Cl
448.388
448.39
Videos