missing translation for 'onlineSavingsMsg'
Learn More

GR 144053 trihydrochloride, Tocris Bioscience™

Glycoprotein IIb/IIIa (integrin αIIbβ3) receptor antagonist. Antithrombotic

Brand:  Tocris (Bio-Techne) 126350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13449013

  • £649.74 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent and selective platelet fibrinogen receptor glycoprotein IIb/IIIa (GpIIb/IIIa) antagonist (IC50 = 37 nM). Orally active and highly effective at inhibiting thrombus formation in vivo.
Specifications

Chemical Identifiers

1215333-48-4
454.821
gr 144053 trihydrochloride, 4-4-4-aminoiminomethyl phenyl-1-piperazinyl-1-piperidineacetic acid trihydrochloride, 2-4-4-4-carbamimidoylphenyl piperazin-1-yl piperidin-1-yl acetic acid trihydrochloride
2-[4-[4-(4-carbamimidoylphenyl)piperazin-1-yl]piperidin-1-yl]acetic acid;trihydrochloride
C18H30Cl3N5O2
YKRNPHOBDOUQTG-UHFFFAOYSA-N
16759148
C1CN(CCC1N2CCN(CC2)C3=CC=C(C=C3)C(=N)N)CC(=O)O.Cl.Cl.Cl

Specifications

50mg
1215333-48-4
gr 144053 trihydrochloride, 4-4-4-aminoiminomethyl phenyl-1-piperazinyl-1-piperidineacetic acid trihydrochloride, 2-4-4-4-carbamimidoylphenyl piperazin-1-yl piperidin-1-yl acetic acid trihydrochloride
C1CN(CCC1N2CCN(CC2)C3=CC=C(C=C3)C(=N)N)CC(=O)O.Cl.Cl.Cl
454.821
454.83
GR 144053 trihydrochloride
C18H30Cl3N5O2
YKRNPHOBDOUQTG-UHFFFAOYSA-N
2-[4-[4-(4-carbamimidoylphenyl)piperazin-1-yl]piperidin-1-yl]acetic acid;trihydrochloride
16759148
>99%
Videos
Documents