Learn More
GR 135531, Tocris Bioscience™
High affinity melatonin MT3 ligand
Brand:  Tocris (Bio-Techne) 089650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13438123
  • £393.92 / 50mg
Estimated Shipment
08-07-2019
Description
Description
Binds with high affinity to MT3 melatonin sites in hamster brain, and low affinity (and no efficacy) at MT1 and MT2 melatonin receptors and thus can be used to discriminate between the melatonin receptor subtypes.
Specifications
Chemical Identifiers
190277-13-5
275.308
5-methoxycarbonylamino-n-acetyltryptamine, 5-mca-nat, 5mca-nat, methyl n-3-2-acetamidoethyl-1h-indol-5-yl carbamate, methyl 3-2-acetylamino ethyl-1h-indol-5-yl carbamate, methyl 3-2-acetylamino ethyl-1h-indol-5-yl carbamate, tocris-0896, d01lau, methyl 3-2-acetamidoethyl-1h-indol-5-ylcarbamate, n-3-2-acetylamino ethyl-1h-indol-5-yl carbamic acid methyl ester
methyl N-[3-(2-acetamidoethyl)-1H-indol-5-yl]carbamate
C14H17N3O3
MPZVHKLZCUEJFO-UHFFFAOYSA-N
4284525
CC(=O)NCCC1=CNC2=C1C=C(C=C2)NC(=O)OC
Specifications
GR 135531
190277-13-5
5-methoxycarbonylamino-n-acetyltryptamine, 5-mca-nat, 5mca-nat, methyl n-3-2-acetamidoethyl-1h-indol-5-yl carbamate, methyl 3-2-acetylamino ethyl-1h-indol-5-yl carbamate, methyl 3-2-acetylamino ethyl-1h-indol-5-yl carbamate, tocris-0896, d01lau, methyl 3-2-acetamidoethyl-1h-indol-5-ylcarbamate, n-3-2-acetylamino ethyl-1h-indol-5-yl carbamic acid methyl ester
CC(=O)NCCC1=CNC2=C1C=C(C=C2)NC(=O)OC
275.308
275.31
50mg
C14H17N3O3
MPZVHKLZCUEJFO-UHFFFAOYSA-N
methyl N-[3-(2-acetamidoethyl)-1H-indol-5-yl]carbamate
4284525
>97%
Videos