Learn More
GR 135531, Tocris Bioscience™
High affinity melatonin MT3 ligand
Brand:  Tocris (Bio-Techne) 0896/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13428123
  • £119.07 / 10mg
Estimated Shipment date
from Supplier 02-04-2020
Description
Description
Binds with high affinity to MT3 melatonin sites in hamster brain, and low affinity (and no efficacy) at MT1 and MT2 melatonin receptors and thus can be used to discriminate between the melatonin receptor subtypes.
Specifications
Chemical Identifiers
190277-13-5
275.308
5-methoxycarbonylamino-n-acetyltryptamine, 5-mca-nat, 5mca-nat, methyl n-3-2-acetamidoethyl-1h-indol-5-yl carbamate, methyl 3-2-acetylamino ethyl-1h-indol-5-yl carbamate, tocris-0896, d01lau, methyl 3-2-acetamidoethyl-1h-indol-5-ylcarbamate, n-3-2-acetylamino ethyl-1h-indol-5-yl carbamic acid methyl ester
methyl N-[3-(2-acetamidoethyl)-1H-indol-5-yl]carbamate
C14H17N3O3
MPZVHKLZCUEJFO-UHFFFAOYSA-N
4284525
CC(=O)NCCC1=CNC2=C1C=C(C=C2)NC(=O)OC
Specifications
10mg
190277-13-5
5-methoxycarbonylamino-n-acetyltryptamine, 5-mca-nat, 5mca-nat, methyl n-3-2-acetamidoethyl-1h-indol-5-yl carbamate, methyl 3-2-acetylamino ethyl-1h-indol-5-yl carbamate, tocris-0896, d01lau, methyl 3-2-acetamidoethyl-1h-indol-5-ylcarbamate, n-3-2-acetylamino ethyl-1h-indol-5-yl carbamic acid methyl ester
CC(=O)NCCC1=CNC2=C1C=C(C=C2)NC(=O)OC
275.308
275.31
GR 135531
C14H17N3O3
MPZVHKLZCUEJFO-UHFFFAOYSA-N
methyl N-[3-(2-acetamidoethyl)-1H-indol-5-yl]carbamate
4284525
>97%