missing translation for 'onlineSavingsMsg'
Learn More

GR 103691, Tocris Bioscience™

Brand:  Tocris (Bio-Techne) 110950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458543

  • £610.05 / 50mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

162408-66-4
485.628
unii-0y0cm1a77l, 4'-acetyl-n-4-4-2-methoxyphenyl-1-piperazinyl butyl-1,1'-biphenyl-4-carboxamide, dsstox_cid_22602, dsstox_rid_80056, dsstox_gsid_42602, 4-4-acetylphenyl-n-4-4-2-methoxyphenyl piperazin-1-yl butyl benzamide, 4'-acetyl-n-4-4-2-methoxyphenyl piperazin-1-yl butyl-1,1'-biphenyl-4-carboxamide, tocris-1109, biomol-nt_000041
4-(4-acetylphenyl)-N-[4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl]benzamide
C30H35N3O3
JARNORYOPMINDY-UHFFFAOYSA-N
4302960
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)NCCCCN3CCN(CC3)C4=CC=CC=C4OC

Specifications

50mg
162408-66-4
unii-0y0cm1a77l, 4'-acetyl-n-4-4-2-methoxyphenyl-1-piperazinyl butyl-1,1'-biphenyl-4-carboxamide, dsstox_cid_22602, dsstox_rid_80056, dsstox_gsid_42602, 4-4-acetylphenyl-n-4-4-2-methoxyphenyl piperazin-1-yl butyl benzamide, 4'-acetyl-n-4-4-2-methoxyphenyl piperazin-1-yl butyl-1,1'-biphenyl-4-carboxamide, tocris-1109, biomol-nt_000041
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)NCCCCN3CCN(CC3)C4=CC=CC=C4OC
485.628
485.63
GR 103691
C30H35N3O3
JARNORYOPMINDY-UHFFFAOYSA-N
4-(4-acetylphenyl)-N-[4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl]benzamide
4302960
>98%
Videos
Documents