Learn More
GPRIN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563465
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ASAPSSAAGR DLIHTPLTMP ANQHTCQSIP GDQPNAITSS MPEDSLMRSQ RTSNREQPEK PSCPVGGVLS SSKDQVSCEF PSPETIQGTV QTPV Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 46%.
May be involved in neurite outgrowth.Specifications
GPRIN3 | |
Polyclonal | |
Unconjugated | |
Gprin3 | |
C030038J10Rik; C730021L23; FLJ42625; G protein-regulated inducer of neurite outgrowth 3; GPRIN family member 3; Gprin3; GRIN3; KIAA2027; RGD1565690 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
285513 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZVF9 | |
Gprin3 | |
Recombinant protein corresponding to Human GPRIN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only