missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRIN2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558256
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QAEPKAAEQL ATTTCHALPP AALLCGMREM REVGAGGCCH ALPATGILAF PKLVASVSES GLQAQHGVKI HCRL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.
May be involved in neurite outgrowth. Nagase T., DNA Res. 5:31-39(1998). The MGC Project Team; Genome Res. 14:2121-2127(2004). Chen L.T., J. Biol. Chem. 274:26931-26938(1999).Specifications
GPRIN2 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60269 | |
GPRIN2 | |
Recombit protein corresponding to Human GPRIN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
GPRIN2 | |
GRIN2, KIAA0514, G protein-regulated inducer of neurite outgrowth 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9721 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only