Learn More
GPR156 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555928
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPDLPEQWQL WPPVPSGCAS LSSQHSYFDT ESSSSDEFFC RCHRPYCEIC FQSSSDSSDS GTSDTDPEPT GGLASWEKLW ARSKPIVNFK DDLKPTLV Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 74%.
GPR156 Polyclonal antibody specifically detects GPR156 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
GPR156 | |
Polyclonal | |
Unconjugated | |
GPR156 | |
G protein-coupled receptor 156; G protein-coupled receptor PGR28; GABABL; GABAB-related G-protein coupled receptor; GPR156; G-protein coupled receptor PGR28; PGR28; Probable G-protein coupled receptor 156 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
165829 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFN8 | |
GPR156 | |
Recombinant protein corresponding to Human GPR156 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only