Learn More
GPNMB Monoclonal Antibody (CTSREVL), eFluor™ 660, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 50-5708-82
143.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: This CTSREVL monoclonal antibody recognizes human GPNMB also known as Osteoactivin, DC-HIL and HGFIN. This type-1 transmembrane glycoprotein was described independently as a bone development factor, tumor related molecule, neurokinin, and immune regulator. Expressed on macrophages and dendritic cells, GPNMB is a ligand for proteoglycan Syndecan-4 (SD-4), which is found on T cells and is known to inhibit their activation and proliferation. Hence, surface expression of GPNMB by myeloid cells may be indicative of their tolerogenic properties. Various cancers express GPNMB, and it is suspected to help in immune evasion. Treatment with IL-10 and various immunosuppressive drugs (e.g., dexamethasone) upregulate GPNMB on macrophages and dendritic cells. GPNMB may also be secreted. The CTSREVL antibody will work on paraformaldehyde-fixed cells. Applications Reported: This CTSREVL antibody has been reported for use in flow cytometric analysis. Applications Tested: This CTSREVL antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal exudate cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
GPNMB Monoclonal antibody specifically detects GPNMB in Mouse samples. It is validated for Flow Cytometry.Specifications
GPNMB | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q99P91 | |
GPNMB | |
FC-tagged full protein expressed in insect cells | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
CTSREVL | |
eFluor 660 | |
GPNMB | |
Dchil; DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; dendritic cell-associated transmembrane protein; glycoprotein (transmembrane) nmb; glycoprotein nmb; glycoprotein nmb-like protein; glycoprotein nonmetastatic melanoma protein B; Gpnmb; hematopoietic growth factor inducible neurokinin-1; Hematopoietic growth factor inducible neurokinin-1 type; Hgfin; ipd; iris pigment dispersion; NMB; Osteoactivin; si:ch211-155k24.6; transmembrane glycoprotein HGFIN; transmembrane glycoprotein NMB; Unknown (protein for MGC:133733); UNQ1725/PRO9925 | |
Rat | |
Affinity chromatography | |
RUO | |
93695 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
For Research Use Only.