missing translation for 'onlineSavingsMsg'
Learn More

GPi 688, Tocris Bioscience™

Allosteric glycogen phosphorylase inhibitor

Brand:  Tocris (Bio-Techne) 396750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13433904

  • £770.28 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Allosteric glycogen phosphorylase inhibitor; acts at the indole site of glycogen phosphorylase. Inhibits glucagon-mediated hyperglycemia in vivo in the rat.
Specifications

Chemical Identifiers

918902-32-6
419.88
3,4-dihydro-2-quinolone dhq derivative, 33, 2-chloro-n-1-2 r ,3-dihydroxypropyl-2-oxo-1,2,3,4-tetrahydroquinolin-3 r,s-yl-6h-thieno 2,3-b pyrrole-5-carboxamide, 2-chloro-n-1-2r-2,3-dihydroxypropyl-1,2,3,4-tetrahydro-2-oxo-3-quinolinyl-6h-thieno 2,3-b pyrrole-5-carboxamide, 2-chloro-n-1-2r-2,3-dihydroxypropyl-2-oxo-1,2,3,4-tetrahydroquinolin-3-yl-6h-thieno 2,3-b pyrrole-5-carboxamide
2-chloro-N-[1-[(2R)-2,3-dihydroxypropyl]-2-oxo-3,4-dihydroquinolin-3-yl]-6H-thieno[2,3-b]pyrrole-5-carboxamide
C19H18ClN3O4S
UICNBXVDHCBKCE-PUODRLBUSA-N
10477191
C1C(C(=O)N(C2=CC=CC=C21)CC(CO)O)NC(=O)C3=CC4=C(N3)SC(=C4)Cl

Specifications

50mg
918902-32-6
3,4-dihydro-2-quinolone dhq derivative, 33, 2-chloro-n-1-2 r ,3-dihydroxypropyl-2-oxo-1,2,3,4-tetrahydroquinolin-3 r,s-yl-6h-thieno 2,3-b pyrrole-5-carboxamide, 2-chloro-n-1-2r-2,3-dihydroxypropyl-1,2,3,4-tetrahydro-2-oxo-3-quinolinyl-6h-thieno 2,3-b pyrrole-5-carboxamide, 2-chloro-n-1-2r-2,3-dihydroxypropyl-2-oxo-1,2,3,4-tetrahydroquinolin-3-yl-6h-thieno 2,3-b pyrrole-5-carboxamide
C1C(C(=O)N(C2=CC=CC=C21)CC(CO)O)NC(=O)C3=CC4=C(N3)SC(=C4)Cl
419.88
419.88
GPi 688
C19H18ClN3O4S
UICNBXVDHCBKCE-PUODRLBUSA-N
2-chloro-N-[1-[(2R)-2,3-dihydroxypropyl]-2-oxo-3,4-dihydroquinolin-3-yl]-6H-thieno[2,3-b]pyrrole-5-carboxamide
10477191
>99%
Videos
Documents