Learn More
GPATCH3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556918
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PNSALIPTDP AAEGQLLSQT SATDVRPLST RDSTPIQTRT CCCVISVRGL AQAQRLIRMY SGRRWLDSHG TWLPGRCLIR RLRLPTEASG LGSFPFKTRK EL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.
GPATCH3 Polyclonal antibody specifically detects GPATCH3 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GPATCH3 | |
Polyclonal | |
Unconjugated | |
GPATCH3 | |
D930035B09Rik; G patch domain containing 3; G patch domain-containing protein 3; Gpatc3; G-patch domain containing 3; GPATCH3; RGD1311216 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63906 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96I76 | |
GPATCH3 | |
Recombinant protein corresponding to Human GPATCH3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only