missing translation for 'onlineSavingsMsg'
Learn More

Gossypol-acetic acid, Thermo Scientific Chemicals

Catalog Number p-7057411
Click to view available options
Quantity:
25 mg
This item is not returnable. View return policy
12542-36-8
C32H34O10
578.614
MFCD00058385
NIOHNDKHQHVLKA-UHFFFAOYSA-N
gossypol-acetic acid, gossypol acetate, gossypol acetic acid, acetate gossypol, --gossypol acetic acid, s-gossypol acetic acid, at-101 acetic acid, gossypol-acetic acid complex
227456
acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O
This item is not returnable. View return policy
12542-36-8
C32H34O10
578.614
MFCD00058385
NIOHNDKHQHVLKA-UHFFFAOYSA-N
gossypol-acetic acid, gossypol acetate, gossypol acetic acid, acetate gossypol, --gossypol acetic acid, s-gossypol acetic acid, at-101 acetic acid, gossypol-acetic acid complex
227456
acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Green
Quantity 25 mg
Formula Weight 578.61
Physical Form Powder
Chemical Name or Material Gossypol-acetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'