Learn More
GOSR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554241
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TGVNDKMAEY TNSAGVPSLN AALMHTLQRH RDILQDYTHE FHKTKANFMA IRERENLMGS VRKDIESYKS GSGVNNRRTE LFLKEHDHLR NSDRLIEETI SIAMATKENM TSQRGMLKSI HSKMNTLANR FPAVNSLIQR INLRKR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
GOSR1 Polyclonal antibody specifically detects GOSR1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GOSR1 | |
Polyclonal | |
Unconjugated | |
GOSR1 | |
28 kDa cis-Golgi SNARE p28; 28 kDa Golgi SNARE protein; AI414660; AI426320; BB145494; cis-Golgi p28 (p28); cis-golgi SNARE; golgi integral membrane protein 2; golgi SNAP receptor complex member 1; Golgi SNARE 28 kDa; GOLIM2; Gos28; GOS-28; GOS28/P28; Gosr1; GOSRI; Gs28; hypothetical protein LOC511030; hypothetical protein LOC550344; I79_013357; p28; RP23-343C18.3; v-SNARE; zgc:112064 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9527 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95249 | |
GOSR1 | |
Recombinant protein corresponding to Human GOSR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only